Learn More
Meso Scale Discovery U-PLEX Human IL-1 ASY
SDP
Catalog No. 5000079679
Encompass
The Meso Scale Discovery U-PLEX Human IL-1alpha Assay Kit is a multiplex immunoassay that measures IL-1alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay can quantify IL-1alpha concentrations between 0 06 pg/mL to 10 000 pg/mL It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and an MSD read buffer The total assay time is 3 5 hours
Catalog No. 50-000-79679 Supplier Meso Scale Discovery Supplier No. K151UNK2
The Meso Scale Discovery U-PLEX Human IL-1alpha Assay Kit is a multiplex immunoassay that measures IL-1alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay can quantify IL-1alpha concentrations between 0 06 pg/mL to 10 000 pg/mL It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and an MSD read buffer The total assay time is 3 5 hours