Learn More
Meso Scale Discovery U-PLEX Human IL-1 ASY
SDP
Catalog No. 5000080536
Encompass
The Meso Scale Discovery U-PLEX Human IL-1alpha Assay Kit is a multiplex immunoassay that measures IL-1alpha in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-80536 Supplier Meso Scale Discovery Supplier No. K151UNK1
The Meso Scale Discovery U-PLEX Human IL-1alpha Assay Kit is a multiplex immunoassay that measures IL-1alpha in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL