Learn More
Meso Scale Discovery U-PLEX Human IL-1 ASYQP
SDP
Catalog No. 5000082024
Encompass
The U-PLEX Human IL-1 Assay (K151UNK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-1 alpha in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications
Catalog No. 50-000-82024 Supplier Meso Scale Discovery Supplier No. K151UNK21
The U-PLEX Human IL-1 Assay (K151UNK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-1 alpha in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications