Learn More
Meso Scale Discovery U-PLEX Human IL-10 Assay, 384-well, K251TZK-2, 0.14-3,700 pg/mL dynamic range, for research use only
SDP
Catalog No. NC2944986
Encompass
U-PLEX Human IL-10 Assay, 384-well, K251TZK-2, 0.14-3,700 pg/mL dynamic range, for research use only
Catalog No. NC2944986 Supplier Meso Scale Discovery Supplier No. K251TZK2
Add to Cart
Add to Cart
Request a Quote
U-PLEX Human IL-10 Assay, 384-well, K251TZK-2, 0.14-3,700 pg/mL dynamic range, for research use only