Learn More
Meso Scale Discovery U-PLEX Human IL-10 ASY
SDP
Catalog No. 5000079000
Encompass
The Meso Scale Discovery U-PLEX Human IL-10 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager and QuickPlex SQ 120 instruments with MULTI-ARRAY technology for sensitive accurate results in 3 5 hours The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG reagents calibrators and an assay diluent The detection limit is 0 04 pg/mL and the quantification limit is 2500 pg/mL
Catalog No. 50-000-79000 Supplier Meso Scale Discovery Supplier No. K151TZK1
The Meso Scale Discovery U-PLEX Human IL-10 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses MSD s SECTOR Imager and QuickPlex SQ 120 instruments with MULTI-ARRAY technology for sensitive accurate results in 3 5 hours The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG reagents calibrators and an assay diluent The detection limit is 0 04 pg/mL and the quantification limit is 2500 pg/mL