Learn More
Meso Scale Discovery U-PLEX Human IL-12p70 ASY
SDP
Catalog No. 5000078571
Encompass
The Meso Scale Discovery U-PLEX Human IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 12 (IL-12) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification of IL-12p70 with detection limits from 0 74 to 2500 pg/mL The kit includes a 96-well plate detection antibodies buffers blocker solutions calibrator diluent and lyophilized calibrator It runs on MSD s SECTOR Imager 6000 for separation-free assays
Catalog No. 50-000-78571 Supplier Meso Scale Discovery Supplier No. K151UAK1
The Meso Scale Discovery U-PLEX Human IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 12 (IL-12) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification of IL-12p70 with detection limits from 0 74 to 2500 pg/mL The kit includes a 96-well plate detection antibodies buffers blocker solutions calibrator diluent and lyophilized calibrator It runs on MSD s SECTOR Imager 6000 for separation-free assays