Learn More
Meso Scale Discovery U-PLEX Human IL-12p70 ASYQP
SDP
Catalog No. 5000078526
Encompass
The U-PLEX Human IL-12p70 Assay (K151UAK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-12p70 in biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications
Catalog No. 50-000-78526 Supplier Meso Scale Discovery Supplier No. K151UAK21
The U-PLEX Human IL-12p70 Assay (K151UAK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-12p70 in biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications