Learn More
Meso Scale Discovery U-PLEX Human IL-13 ASY
SDP
Catalog No. 5000079690
Encompass
The Meso Scale Discovery U-PLEX Human IL-13 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-13 in human serum plasma and cell culture supernatants It uses the SECTOR Imager 6000 for high sensitivity and a wide dynamic range The kit includes a 96-well plate with capture antibody detection antibody with MSD SULFO-TAG calibrators and sample diluent It has a detection range of 0 04 to 10 000 pg/mL and high specificity with no cross-reactivity to other cytokines providing all necessary reagents for the assay
Catalog No. 50-000-79690 Supplier Meso Scale Discovery Supplier No. K151UBK1
The Meso Scale Discovery U-PLEX Human IL-13 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-13 in human serum plasma and cell culture supernatants It uses the SECTOR Imager 6000 for high sensitivity and a wide dynamic range The kit includes a 96-well plate with capture antibody detection antibody with MSD SULFO-TAG calibrators and sample diluent It has a detection range of 0 04 to 10 000 pg/mL and high specificity with no cross-reactivity to other cytokines providing all necessary reagents for the assay