Learn More
Meso Scale Discovery U-PLEX Human IL-13 ASY384-well
SDP
Catalog No. 5000081063
Encompass
The Meso Scale Discovery U-PLEX Human IL-13 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-13 in human serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are read on MSD s SECTOR Imager and analyzed with Discovery Workbench software Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL
Catalog No. 50-000-81063 Supplier Meso Scale Discovery Supplier No. K251UBK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-13 Assay Kit is an electrochemiluminescent immunoassay for measuring IL-13 in human serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are read on MSD s SECTOR Imager and analyzed with Discovery Workbench software Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL