Learn More
Meso Scale Discovery U-PLEX Human IL-15 ASY 25 PL
SDP
Catalog No. 5000079733
Encompass
The Meso Scale Discovery U-PLEX Human IL-15 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 64 pg/mL and the assay range is up to 10 000 pg/mL
Catalog No. 50-000-79733 Supplier Meso Scale Discovery Supplier No. K151URK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-15 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 64 pg/mL and the assay range is up to 10 000 pg/mL