Learn More
Meso Scale Discovery U-PLEX Human IL-15 ASY
SDP
Catalog No. 5000079730
Encompass
The Meso Scale Discovery U-PLEX Human IL-15 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 15 (IL-15) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer for signal detection on MSD platforms Sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL for IL-15
Catalog No. 50-000-79730 Supplier Meso Scale Discovery Supplier No. K151URK1
The Meso Scale Discovery U-PLEX Human IL-15 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 15 (IL-15) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer for signal detection on MSD platforms Sensitivity is 0 3 pg/mL with an upper quantification limit of 2500 pg/mL for IL-15