Learn More
Meso Scale Discovery U-PLEX Human IL-15 ASY384-well
SDP
Catalog No. 5000080896
Encompass
The Meso Scale Discovery U-PLEX Human IL-15 Assay Kit is an electrochemiluminescent immunoassay for quantifying human interleukin 15 (IL-15) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive IL-15 quantitation with detection limits from 0 49 to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range offering a simple rapid and robust IL-15 measurement method
Catalog No. 50-000-80896 Supplier Meso Scale Discovery Supplier No. K251URK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-15 Assay Kit is an electrochemiluminescent immunoassay for quantifying human interleukin 15 (IL-15) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive IL-15 quantitation with detection limits from 0 49 to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range offering a simple rapid and robust IL-15 measurement method