Learn More
Meso Scale Discovery U-PLEX Human IL-16 ASY 25 PL
SDP
Catalog No. 5000078540
Encompass
The Meso Scale Discovery U-PLEX Human IL-16 Assay Kit is a multiplex immunoassay for measuring IL-16 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity a 5-log dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls for a standard curve It has high specificity and under 1% cross-reactivity with other cytokines performing up to 80 tests in 3 5 hours
Catalog No. 50-000-78540 Supplier Meso Scale Discovery Supplier No. K151USK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-16 Assay Kit is a multiplex immunoassay for measuring IL-16 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity a 5-log dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls for a standard curve It has high specificity and under 1% cross-reactivity with other cytokines performing up to 80 tests in 3 5 hours