Learn More
Meso Scale Discovery U-PLEX Human IL-16 ASY
SDP
Catalog No. 5000079148
Encompass
The Meso Scale Discovery U-PLEX Human IL-16 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-16 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer for analysis on MSD instruments The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79148 Supplier Meso Scale Discovery Supplier No. K151USK2
The Meso Scale Discovery U-PLEX Human IL-16 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-16 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer for analysis on MSD instruments The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL