Learn More
Meso Scale Discovery U-PLEX Human IL-16 ASY384-well
SDP
Catalog No. 5000080660
Encompass
The Meso Scale Discovery U-PLEX Human IL-16 384-Well Assay Kit is a multiplex immunoassay for measuring IL-16 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls for a standard curve It has high specificity and 1% cross-reactivity with other cytokines requiring 50 L per well and allowing up to 40 tests in duplicate
Catalog No. 50-000-80660 Supplier Meso Scale Discovery Supplier No. K251USK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-16 384-Well Assay Kit is a multiplex immunoassay for measuring IL-16 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls for a standard curve It has high specificity and 1% cross-reactivity with other cytokines requiring 50 L per well and allowing up to 40 tests in duplicate