Learn More
Meso Scale Discovery U-PLEX Human IL-17A ASYQP
SDP
Catalog No. 5000078248
Encompass
The U-PLEX Human IL-17A Assay (K151UTK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17A levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-17A detection supporting research in inflammation autoimmune diseases and immunology studies The kit provides reliable reproducible results with minimal sample volume requirements and is compatible with various sample types including serum plasma and cell culture supernatants
Catalog No. 50-000-78248 Supplier Meso Scale Discovery Supplier No. K151UTK21
The U-PLEX Human IL-17A Assay (K151UTK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17A levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-17A detection supporting research in inflammation autoimmune diseases and immunology studies The kit provides reliable reproducible results with minimal sample volume requirements and is compatible with various sample types including serum plasma and cell culture supernatants