Learn More
Meso Scale Discovery U-PLEX Human IL-17A/F ASY
SDP
Catalog No. 5000079738
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A/F Assay Kit is a multiplex immunoassay that measures human interleukin 17A and interleukin 17F in cell culture supernatants serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It can detect IL-17A in a range of 0 06-1500 pg/mL and IL-17F in a range of 0 10-1500 pg/mL
Catalog No. 50-000-79738 Supplier Meso Scale Discovery Supplier No. K151VYK2
The Meso Scale Discovery U-PLEX Human IL-17A/F Assay Kit is a multiplex immunoassay that measures human interleukin 17A and interleukin 17F in cell culture supernatants serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It can detect IL-17A in a range of 0 06-1500 pg/mL and IL-17F in a range of 0 10-1500 pg/mL