Learn More
Meso Scale Discovery U-PLEX Human IL-17A/F ASY
SDP
Catalog No. 5000081838
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A/F Assay Kit is a multiplex immunoassay that measures IL-17A and IL-17F in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay takes 3 5 hours to complete and includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator and control solutions It allows simultaneous quantification of IL-17A and IL-17F from a single sample for understanding Th17 responses
Catalog No. 50-000-81838 Supplier Meso Scale Discovery Supplier No. K151VYK1
The Meso Scale Discovery U-PLEX Human IL-17A/F Assay Kit is a multiplex immunoassay that measures IL-17A and IL-17F in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay takes 3 5 hours to complete and includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator and control solutions It allows simultaneous quantification of IL-17A and IL-17F from a single sample for understanding Th17 responses