Learn More
Meso Scale Discovery U-PLEX Human IL-17B ASY
SDP
Catalog No. 5000078751
Encompass
The Meso Scale Discovery U-PLEX Human IL-17B Assay Kit is a multiplex immunoassay that measures IL-17B levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent This assay allows for the accurate and reliable quantification of IL-17B levels in approximately 3 5 hours
Catalog No. 50-000-78751 Supplier Meso Scale Discovery Supplier No. K151XNK1
The Meso Scale Discovery U-PLEX Human IL-17B Assay Kit is a multiplex immunoassay that measures IL-17B levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent This assay allows for the accurate and reliable quantification of IL-17B levels in approximately 3 5 hours