Learn More
Meso Scale Discovery U-PLEX Human IL-17B ASY
SDP
Catalog No. 5000080458
Encompass
The Meso Scale Discovery U-PLEX Human IL-17B Assay Kit quantifies human interleukin 17B (IL-17B) in serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive cross-reactivity-free detection It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s QuickPlex SQ 120 offering a 4 5-hour protocol for accurate IL-17B quantification
Catalog No. 50-000-80458 Supplier Meso Scale Discovery Supplier No. K151XNK2
The Meso Scale Discovery U-PLEX Human IL-17B Assay Kit quantifies human interleukin 17B (IL-17B) in serum plasma and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive cross-reactivity-free detection It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s QuickPlex SQ 120 offering a 4 5-hour protocol for accurate IL-17B quantification