Learn More
Meso Scale Discovery U-PLEX Human IL-17B ASYQP
SDP
Catalog No. 5000079706
Encompass
The U-PLEX Human IL-17B Assay (K151XNK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17B levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-17B detection supporting research into inflammatory and immune responses The kit provides reliable reproducible results for clinical and research applications studying cytokine signaling pathways
Catalog No. 50-000-79706 Supplier Meso Scale Discovery Supplier No. K151XNK22
The U-PLEX Human IL-17B Assay (K151XNK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17B levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-17B detection supporting research into inflammatory and immune responses The kit provides reliable reproducible results for clinical and research applications studying cytokine signaling pathways