Learn More
Meso Scale Discovery U-PLEX Human IL-17C ASY
SDP
Catalog No. 5000081477
Encompass
The Meso Scale Discovery U-PLEX Human IL-17C Assay Kit is designed for the quantitative measurement of human interleukin 17C (IL-17C) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection technology The assay uses a plate that is pre-coated with capture antibodies on distinct spots allowing for the measurement of IL-17C and other biomarkers in the same sample The sensitivity of the assay is excellent with a lower limit of detection of 0 08 pg/mL The assay takes approximately 3 5 hours to complete Overall the U-PLEX Human IL-17C Assay Kit provides a sensitive reproducible and flexible way to quantitatively measure human IL-17C
Catalog No. 50-000-81477 Supplier Meso Scale Discovery Supplier No. K151WJK2
The Meso Scale Discovery U-PLEX Human IL-17C Assay Kit is designed for the quantitative measurement of human interleukin 17C (IL-17C) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection technology The assay uses a plate that is pre-coated with capture antibodies on distinct spots allowing for the measurement of IL-17C and other biomarkers in the same sample The sensitivity of the assay is excellent with a lower limit of detection of 0 08 pg/mL The assay takes approximately 3 5 hours to complete Overall the U-PLEX Human IL-17C Assay Kit provides a sensitive reproducible and flexible way to quantitatively measure human IL-17C