Learn More
Meso Scale Discovery U-PLEX Human IL-17C ASYQP
SDP
Catalog No. 5000082396
Encompass
The U-PLEX Human IL-17C Assay (K151WJK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-17C protein levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms to deliver sensitive precise detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications
Catalog No. 50-000-82396 Supplier Meso Scale Discovery Supplier No. K151WJK22
The U-PLEX Human IL-17C Assay (K151WJK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-17C protein levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms to deliver sensitive precise detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable cytokine analysis in research applications