Learn More
Meso Scale Discovery U-PLEX Human IL-17D ASY
SDP
Catalog No. 5000078750
Encompass
The Meso Scale Discovery U-PLEX Human IL-17D Assay Kit is a multiplex immunoassay for measuring IL-17D in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and diluents It quantifies IL-17D with minimal hands-on time using an MSD instrument providing a simple rapid method to measure IL-17D important for immune regulation
Catalog No. 50-000-78750 Supplier Meso Scale Discovery Supplier No. K151XOK2
The Meso Scale Discovery U-PLEX Human IL-17D Assay Kit is a multiplex immunoassay for measuring IL-17D in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and diluents It quantifies IL-17D with minimal hands-on time using an MSD instrument providing a simple rapid method to measure IL-17D important for immune regulation