Learn More
Meso Scale Discovery U-PLEX Human IL-17D ASY
SDP
Catalog No. 5000079209
Encompass
The Meso Scale Discovery U-PLEX Human IL-17D Assay Kit is a multiplex immunoassay that measures IL-17D levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents needed to perform the assay The kit can quantitatively measure IL-17D levels with high precision and accuracy It is for research use only
Catalog No. 50-000-79209 Supplier Meso Scale Discovery Supplier No. K151XOK1
The Meso Scale Discovery U-PLEX Human IL-17D Assay Kit is a multiplex immunoassay that measures IL-17D levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents needed to perform the assay The kit can quantitatively measure IL-17D levels with high precision and accuracy It is for research use only