Learn More
Meso Scale Discovery U-PLEX Human IL-17E/IL-25 ASY
SDP
Catalog No. 5000079740
Encompass
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 Assay Kit is a multiplex immunoassay that measures IL-17E and IL-25 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Sample Diluent and Calibrator The kit has a typical sensitivity of 0 04 pg/mL for IL-17E and 0 03 pg/mL for IL-25
Catalog No. 50-000-79740 Supplier Meso Scale Discovery Supplier No. K151VZK2
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 Assay Kit is a multiplex immunoassay that measures IL-17E and IL-25 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Sample Diluent and Calibrator The kit has a typical sensitivity of 0 04 pg/mL for IL-17E and 0 03 pg/mL for IL-25