Learn More
Meso Scale Discovery U-PLEX Human IL-17E/IL-25 ASY
SDP
Catalog No. 5000081731
Encompass
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 Assay Kit is designed for the quantitative measurement of human IL-17E and IL-25 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD Streptavidin-coated plates and U-PLEX Linker to capture and detect the analytes in a sandwich immunoassay format The kit includes a plate calibrator detection antibody read buffer 10X wash buffer sample diluent and complete components required to run the assay The sensitivity of the assay is sub-picogram per mL levels The assay provides a reproducible and robust method for measuring IL-17E and IL-25 levels in biological samples
Catalog No. 50-000-81731 Supplier Meso Scale Discovery Supplier No. K151VZK1
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 Assay Kit is designed for the quantitative measurement of human IL-17E and IL-25 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD GOLD Streptavidin-coated plates and U-PLEX Linker to capture and detect the analytes in a sandwich immunoassay format The kit includes a plate calibrator detection antibody read buffer 10X wash buffer sample diluent and complete components required to run the assay The sensitivity of the assay is sub-picogram per mL levels The assay provides a reproducible and robust method for measuring IL-17E and IL-25 levels in biological samples