Learn More
Meso Scale Discovery U-PLEX Human IL-17F ASY
SDP
Catalog No. 5000078722
Encompass
The Meso Scale Discovery U-PLEX Human IL-17F Assay Kit is a multiplex immunoassay that measures human IL-17F levels in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-78722 Supplier Meso Scale Discovery Supplier No. K151WAK1
The Meso Scale Discovery U-PLEX Human IL-17F Assay Kit is a multiplex immunoassay that measures human IL-17F levels in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL