Learn More
Meso Scale Discovery U-PLEX Human IL-18 Assay, Single-analyte, 5 plates minimum order, 96-well, LLOD 0.5 pg/mL, Dynamic range 0.5-14,000 pg/mL, Serum, Plasma, Cell Culture Supernatant
SDP
Catalog No. NC2232684
Encompass
U-PLEX Human IL-18 Assay, Single-analyte, 5 plates minimum order, 96-well, LLOD 0.5 pg/mL, Dynamic range 0.5-14,000 pg/mL, Serum, Plasma, Cell Culture Supernatant
This item is not returnable. View return policy
Catalog No. NC2232684 Supplier Meso Scale Discovery Supplier No. K151VJK2
U-PLEX Human IL-18 Assay, Single-analyte, 5 plates minimum order, 96-well, LLOD 0.5 pg/mL, Dynamic range 0.5-14,000 pg/mL, Serum, Plasma, Cell Culture Supernatant