Learn More
Meso Scale Discovery U-PLEX Human IL-18 ASY 25 PL
SDP
Catalog No. 5000081727
Encompass
The Meso Scale Discovery U-PLEX Human IL-18 Assay Kit is a multiplex immunoassay that measures human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent This assay allows for the quantitative measurement of IL-18 levels with high precision and accuracy
Catalog No. 50-000-81727 Supplier Meso Scale Discovery Supplier No. K151VJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-18 Assay Kit is a multiplex immunoassay that measures human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent This assay allows for the quantitative measurement of IL-18 levels with high precision and accuracy