Learn More
Meso Scale Discovery U-PLEX Human IL-18 ASY
SDP
Catalog No. 5000081836
Encompass
The Meso Scale Discovery U-PLEX Human IL-18 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-18 with a detection range of 0 05-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator This assay allows for the accurate quantification of IL-18 levels in human samples
Catalog No. 50-000-81836 Supplier Meso Scale Discovery Supplier No. K151VJK1
The Meso Scale Discovery U-PLEX Human IL-18 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-18 with a detection range of 0 05-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator This assay allows for the accurate quantification of IL-18 levels in human samples