Learn More
Meso Scale Discovery U-PLEX Human IL-18 ASY384-well
SDP
Catalog No. 5000080512
Encompass
The Meso Scale Discovery U-PLEX Human IL-18 Assay Kit is a multiplex immunoassay that measures IL-18 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80512 Supplier Meso Scale Discovery Supplier No. K251VJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-18 Assay Kit is a multiplex immunoassay that measures IL-18 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 10 000 pg/mL