Learn More
Meso Scale Discovery U-PLEX Human IL-18 ASYQP
SDP
Catalog No. 5000081437
Encompass
The U-PLEX Human IL-18 Assay (K151VJK) is a multiplex immunoassay designed to quantitatively measure interleukin-18 levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-18 detection enabling researchers to study inflammatory responses and immune system function The assay utilizes MSD s U-PLEX platform technology for reliable reproducible results in clinical and research applications
Catalog No. 50-000-81437 Supplier Meso Scale Discovery Supplier No. K151VJK21
The U-PLEX Human IL-18 Assay (K151VJK) is a multiplex immunoassay designed to quantitatively measure interleukin-18 levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-18 detection enabling researchers to study inflammatory responses and immune system function The assay utilizes MSD s U-PLEX platform technology for reliable reproducible results in clinical and research applications