Learn More
Meso Scale Discovery U-PLEX Human IL-18 ASYQP
SDP
Catalog No. 5000081918
Encompass
The U-PLEX Human IL-18 Assay (K151VJK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-18 protein levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-18 quantification in research applications studying inflammation and immune responses
Catalog No. 50-000-81918 Supplier Meso Scale Discovery Supplier No. K151VJK22
The U-PLEX Human IL-18 Assay (K151VJK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-18 protein levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-18 quantification in research applications studying inflammation and immune responses