Learn More
Meso Scale Discovery U-PLEX Human IL-1B ASY 25 PL
SDP
Catalog No. 5000080500
Encompass
The Meso Scale Discovery U-PLEX Human IL-1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a detection range of 0 05-1000 pg/mL and a lower limit of detection of 0 03 pg/mL The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and assay diluent The assay can be run on MSD s SECTOR Imager instrument platforms and data analyzed using MSD Discovery Workbench software
Catalog No. 50-000-80500 Supplier Meso Scale Discovery Supplier No. K151TUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a detection range of 0 05-1000 pg/mL and a lower limit of detection of 0 03 pg/mL The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and assay diluent The assay can be run on MSD s SECTOR Imager instrument platforms and data analyzed using MSD Discovery Workbench software