Learn More
Meso Scale Discovery U-PLEX Human IL-2 ASY 25 PL
SDP
Catalog No. 5000079671
Encompass
The Meso Scale Discovery U-PLEX Human IL-2 Assay Kit is a sensitive immunoassay for detecting and quantifying human interleukin 2 (IL-2) in various biological matrices It uses MSD s electrochemiluminescent detection technology and works with the QuickPlex SQ 120 instrument The assay detects IL-2 in the range of 0 06-1500 pg/mL with minimal cross-reactivity Each kit assays 40 samples in duplicate on a 96-well plate in about 3 hours supporting research in immunology oncology and vaccine development
Catalog No. 50-000-79671 Supplier Meso Scale Discovery Supplier No. K151TVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-2 Assay Kit is a sensitive immunoassay for detecting and quantifying human interleukin 2 (IL-2) in various biological matrices It uses MSD s electrochemiluminescent detection technology and works with the QuickPlex SQ 120 instrument The assay detects IL-2 in the range of 0 06-1500 pg/mL with minimal cross-reactivity Each kit assays 40 samples in duplicate on a 96-well plate in about 3 hours supporting research in immunology oncology and vaccine development