Learn More
Meso Scale Discovery U-PLEX Human IL-2 ASY
SDP
Catalog No. 5000081988
Encompass
The Meso Scale Discovery U-PLEX Human IL-2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 2 (IL-2) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 06 pg/mL
Catalog No. 50-000-81988 Supplier Meso Scale Discovery Supplier No. K151TVK1
The Meso Scale Discovery U-PLEX Human IL-2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 2 (IL-2) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 06 pg/mL