Learn More
Meso Scale Discovery U-PLEX Human IL-21 ASY
SDP
Catalog No. 5000079753
Encompass
The Meso Scale Discovery U-PLEX Human IL-21 Assay Kit is a sensitive immunoassay for detecting and quantifying human interleukin 21 (IL-21) in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator and Control It enables accurate reproducible and sensitive IL-21 measurement within 90 minutes
Catalog No. 50-000-79753 Supplier Meso Scale Discovery Supplier No. K151WBK1
The Meso Scale Discovery U-PLEX Human IL-21 Assay Kit is a sensitive immunoassay for detecting and quantifying human interleukin 21 (IL-21) in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator and Control It enables accurate reproducible and sensitive IL-21 measurement within 90 minutes