Learn More
Meso Scale Discovery U-PLEX Human IL-21 ASY384-well
SDP
Catalog No. 5000080649
Encompass
The Meso Scale Discovery U-PLEX Human IL-21 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human IL-21 in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection down to 0 04 pg/mL The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent The assay takes about 3 5 hours and requires an MSD instrument like the MESO QuickPlex SQ 120 It can perform up to 380 tests
Catalog No. 50-000-80649 Supplier Meso Scale Discovery Supplier No. K251WBK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-21 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human IL-21 in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection down to 0 04 pg/mL The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent The assay takes about 3 5 hours and requires an MSD instrument like the MESO QuickPlex SQ 120 It can perform up to 380 tests