Learn More
Meso Scale Discovery U-PLEX Human IL-21 ASYQP
SDP
Catalog No. 5000081858
Encompass
The MSD U-PLEX Human IL-21 Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of interleukin-21 in human biological samples This singleplex format allows focused detection of IL-21 a cytokine involved in immune regulation and T-cell function The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology and inflammation studies
Catalog No. 50-000-81858 Supplier Meso Scale Discovery Supplier No. K151WBK22
The MSD U-PLEX Human IL-21 Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of interleukin-21 in human biological samples This singleplex format allows focused detection of IL-21 a cytokine involved in immune regulation and T-cell function The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology and inflammation studies