Learn More
Meso Scale Discovery U-PLEX Human IL-22 ASY384-well
SDP
Catalog No. 5000080720
Encompass
The Meso Scale Discovery U-PLEX Human IL-22 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 22 (IL-22) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection with a detection limit of 0 06 pg/mL and quantification up to 1500 pg/mL The kit includes a 384-well plate with IL-22 capture antibody detection antibody calibrators and diluent The assay takes 3 5 hours
Catalog No. 50-000-80720 Supplier Meso Scale Discovery Supplier No. K251WIK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-22 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 22 (IL-22) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection with a detection limit of 0 06 pg/mL and quantification up to 1500 pg/mL The kit includes a 384-well plate with IL-22 capture antibody detection antibody calibrators and diluent The assay takes 3 5 hours