Learn More
Meso Scale Discovery U-PLEX Human IL-23 ASYQP
SDP
Catalog No. 5000081899
Encompass
The U-PLEX Human IL-23 Assay (K151WGK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-23 in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-23 quantification in research applications
Catalog No. 50-000-81899 Supplier Meso Scale Discovery Supplier No. K151WGK21
The U-PLEX Human IL-23 Assay (K151WGK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-23 in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-23 quantification in research applications