Learn More
Meso Scale Discovery U-PLEX Human IL-27 ASY 25 PL
SDP
Catalog No. 5000079635
Encompass
The Meso Scale Discovery U-PLEX Human IL-27 Assay Kit is a multiplex immunoassay for measuring IL-27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG 10X Read Buffer Sample Diluent and calibrators (3 2 - 10 000 pg/mL) It has high specificity and 1% cross-reactivity with other cytokines allowing simultaneous measurement of IL-27 and other biomarkers Validated for research use only
Catalog No. 50-000-79635 Supplier Meso Scale Discovery Supplier No. K151WCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-27 Assay Kit is a multiplex immunoassay for measuring IL-27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG 10X Read Buffer Sample Diluent and calibrators (3 2 - 10 000 pg/mL) It has high specificity and 1% cross-reactivity with other cytokines allowing simultaneous measurement of IL-27 and other biomarkers Validated for research use only