Learn More
Meso Scale Discovery U-PLEX Human IL-27 ASY
SDP
Catalog No. 5000079754
Encompass
The Meso Scale Discovery U-PLEX Human IL-27 Assay Kit is a multiplex immunoassay for measuring IL-27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It requires 25 L of sample and can perform up to 80 tests in duplicate enabling accurate and rapid IL-27 quantification
Catalog No. 50-000-79754 Supplier Meso Scale Discovery Supplier No. K151WCK2
The Meso Scale Discovery U-PLEX Human IL-27 Assay Kit is a multiplex immunoassay for measuring IL-27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It requires 25 L of sample and can perform up to 80 tests in duplicate enabling accurate and rapid IL-27 quantification