Learn More
Meso Scale Discovery U-PLEX Human IL-27 ASY384-well
SDP
Catalog No. 5000079417
Encompass
The U-PLEX Human IL-27 (384-well Assay) is an immunoassay kit for the quantitative measurement of IL-27 in human serum plasma and cell culture supernatants It utilizes Meso Scale Discovery electrochemiluminescence technology and provides a 384-well plate coated with capture antibodies detection antibodies and calibrators The kit has a detection range of 1 95 - 2500 pg/mL and includes all necessary reagents It requires 2 5 hours total assay time and 25 L sample size This assay allows high-throughput sensitive and reproducible quantification of IL-27
Catalog No. 50-000-79417 Supplier Meso Scale Discovery Supplier No. K251WCK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-27 (384-well Assay) is an immunoassay kit for the quantitative measurement of IL-27 in human serum plasma and cell culture supernatants It utilizes Meso Scale Discovery electrochemiluminescence technology and provides a 384-well plate coated with capture antibodies detection antibodies and calibrators The kit has a detection range of 1 95 - 2500 pg/mL and includes all necessary reagents It requires 2 5 hours total assay time and 25 L sample size This assay allows high-throughput sensitive and reproducible quantification of IL-27