Learn More
Meso Scale Discovery U-PLEX Human IL-27 ASYQP
SDP
Catalog No. 5000078732
Encompass
The MSD U-PLEX Human IL-27 Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of interleukin-27 in human biological samples This singleplex format allows focused detection of IL-27 a cytokine involved in immune regulation and inflammatory responses The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology and inflammation studies
Catalog No. 50-000-78732 Supplier Meso Scale Discovery Supplier No. K151WCK22
The MSD U-PLEX Human IL-27 Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of interleukin-27 in human biological samples This singleplex format allows focused detection of IL-27 a cytokine involved in immune regulation and inflammatory responses The U-PLEX platform offers high sensitivity wide dynamic range and minimal sample volume requirements making it suitable for research applications in immunology and inflammation studies