Learn More
Meso Scale Discovery U-PLEX Human IL-2R ASY
SDP
Catalog No. 5000080456
Encompass
The Meso Scale Discovery U-PLEX Human IL-2Ralpha Assay Kit is a multiplex immunoassay that measures human interleukin-2 receptor alpha (IL-2Ralpha) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and assay diluent The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-80456 Supplier Meso Scale Discovery Supplier No. K151XGK1
The Meso Scale Discovery U-PLEX Human IL-2Ralpha Assay Kit is a multiplex immunoassay that measures human interleukin-2 receptor alpha (IL-2Ralpha) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with minimal matrix interference The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators and assay diluent The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 2500 pg/mL