Learn More
Meso Scale Discovery U-PLEX Human IL-2R ASYQP
SDP
Catalog No. 5000080457
Encompass
The U-PLEX Human IL-2R Assay (K151XGK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-2 receptor alpha (CD25) in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for research applications studying immune system activation T-cell responses and inflammatory conditions The kit provides reliable reproducible results for clinical and translational research requiring precise IL-2R detection and quantification
Catalog No. 50-000-80457 Supplier Meso Scale Discovery Supplier No. K151XGK21
The U-PLEX Human IL-2R Assay (K151XGK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-2 receptor alpha (CD25) in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for research applications studying immune system activation T-cell responses and inflammatory conditions The kit provides reliable reproducible results for clinical and translational research requiring precise IL-2R detection and quantification