Learn More
Meso Scale Discovery U-PLEX Human IL-3 ASY 25 PL
SDP
Catalog No. 5000079792
Encompass
The Meso Scale Discovery U-PLEX Human IL-3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-3 with a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The total assay time is 3 5 hours
Catalog No. 50-000-79792 Supplier Meso Scale Discovery Supplier No. K151XMK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-3 with a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The total assay time is 3 5 hours