Learn More
Meso Scale Discovery U-PLEX Human IL-3 ASY
SDP
Catalog No. 5000078752
Encompass
The Meso Scale Discovery U-PLEX Human IL-3 Assay Kit is a multiplex immunoassay for measuring human interleukin 3 (IL-3) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument like MESO QuickPlex SQ 120 aiding in understanding immune responses by quantifying IL-3 levels
Catalog No. 50-000-78752 Supplier Meso Scale Discovery Supplier No. K151XMK2
The Meso Scale Discovery U-PLEX Human IL-3 Assay Kit is a multiplex immunoassay for measuring human interleukin 3 (IL-3) in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument like MESO QuickPlex SQ 120 aiding in understanding immune responses by quantifying IL-3 levels